Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Methyl-2-pyridyl)acetamide 98.0+%, TCI America™
SDP

Catalog No. M27875G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27875G 5 g
M27871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27875G Supplier TCI America Supplier No. M27875G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5327-32-2
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00234432
InChI Key QGZHGSGLCZEGHA-UHFFFAOYSA-N
Synonym 2-Acetamido-4-methylpyridine
PubChem CID 220485
IUPAC Name N-(4-methylpyridin-2-yl)acetamide
SMILES CC1=CC(=NC=C1)NC(=O)C

Specifications

Melting Point 104°C
Color White
Quantity 5 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Methyl-2-pyridyl)acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.