Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Nitrobenzoyl)-beta-alanine 98.0+%, TCI America™
SDP

Catalog No. N09425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09425G 5 g
N094225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09425G Supplier TCI America Supplier No. N09425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 59642-21-6 | C10H9N2O5 | 237.19 g/mol

Chemical Identifiers

CAS 59642-21-6
Molecular Formula C10H9N2O5
Molecular Weight (g/mol) 237.19
MDL Number MFCD00009794
InChI Key PDTLZWITKYGYDN-UHFFFAOYSA-M
Synonym 3-(4-Nitrobenzamido)propionic Acid
PubChem CID 591374
IUPAC Name 3-[(4-nitrophenyl)formamido]propanoate
SMILES [O-]C(=O)CCNC(=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 165°C
Color Yellow
Quantity 5 g
Formula Weight 238.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(4-Nitrobenzoyl)-beta-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.