Alfa Aesar™ N-(4-Pentynyl)phthalimide, 97%

Manufacturer: Alfa Aesar™ H5343206

Quantity 5g

 View more versions of this product

Catalog No. AAH5343206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6097-07-0
    Molecular Formula C13H11NO2
    Formula Weight 213.24
    MDL Number MFCD06798110
    Synonym 1-Phthalimido-4-pentyne
    Chemical Name or Material N-(4-Pentynyl)phthalimide

    Specifications

    Formula Weight 213.24
    Molecular Formula C13H11NO2
    MDL Number MFCD06798110
    CAS 6097-07-0
    Synonym 1-Phthalimido-4-pentyne
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H319-H317
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P363-P321-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 87° to 91°C
    Quantity 5g
    Chemical Name or Material N-(4-Pentynyl)phthalimide
    Assay 97%

    Description

    Applications
    N-(4-Pentynyl)phthalimide is used as pharmaceutical intermediate.

    Notes
    Stable under recommended storage conditions. Store away from oxidizing agents.

    GHS H Statement
    H319-H317
    Causes serious eye irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P305+P351+P338-P363-P321-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit