Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(4-phenyl-1,2,3,4-tetrahydronaphthalen-2-yl)propanamide | 134865-74-0 | MFCD00901545 | >99.0% | 279.38 g/mol | C19H21NO | 100 MG
SDP

Catalog No. 5000214684
Encompass_Preferred

4-P-PDOT is an analytical standard of a selective melatonin MT2 receptor antagonist used as a reference compound in pharmacology and analytical research. It is supplied as a high-purity solid for characterization, assay development, and receptor pharmacology studies.

  • Analytical standard for research and assay calibration.
  • Selective MT2 melatonin receptor antagonist.
  • High purity (>99.0%).
  • Molecular formula C19H21NO.
  • Molecular weight 279.38 g/mol.
  • Provided as a 100 MG solid.
  • CAS number 134865-74-0.

Catalog No. 50-002-14684 Supplier Medchemexpress LLC Supplier No. HY100609100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-P-PDOT is an analytical standard of a selective melatonin MT2 receptor antagonist used as a reference compound in pharmacology and analytical research. It is supplied as a high-purity solid for characterization, assay development, and receptor pharmacology studies.

  • Analytical standard for research and assay calibration.
  • Selective MT2 melatonin receptor antagonist.
  • High purity (>99.0%).
  • Molecular formula C19H21NO.
  • Molecular weight 279.38 g/mol.
  • Provided as a 100 MG solid.
  • CAS number 134865-74-0.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.