Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(4-phenyl-1,2,3,4-tetrahydronaphthalen-2-yl)propanamide | 134865-74-0 | MFCD00901545 | >99.0% | 279.38 g/mol | C19H21NO | 50 MG
SDP

Catalog No. 5000213752
Encompass_Preferred

4-P-PDOT is a potent, selective melatonin MT2 receptor antagonist used as an analytical standard and research reagent. It exhibits >300-fold selectivity for MT2 over MT1 and is supplied in small quantities suitable for biochemical and pharmacological studies; consult the Certificate of Analysis for purity and storage.

  • Potent and selective MT2 antagonist.
  • High receptor selectivity (>300-fold) for MT2 versus MT1.
  • Intended for use as an analytical standard and research reagent.
  • Available in small packaging sizes, including 50 mg.
  • Molecular formula C19H21NO and molecular weight 279.38 g/mol.
  • CAS number 134865-74-0 for unambiguous identification.

Catalog No. 50-002-13752 Supplier Medchemexpress LLC Supplier No. HY10060950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-P-PDOT is a potent, selective melatonin MT2 receptor antagonist used as an analytical standard and research reagent. It exhibits >300-fold selectivity for MT2 over MT1 and is supplied in small quantities suitable for biochemical and pharmacological studies; consult the Certificate of Analysis for purity and storage.

  • Potent and selective MT2 antagonist.
  • High receptor selectivity (>300-fold) for MT2 versus MT1.
  • Intended for use as an analytical standard and research reagent.
  • Available in small packaging sizes, including 50 mg.
  • Molecular formula C19H21NO and molecular weight 279.38 g/mol.
  • CAS number 134865-74-0 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.