Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(4-Piperidin-1-ylbenzyl)cyclopropanamine, ≥95%, Thermo Scientific™

Catalog No. CC33958CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC33958CB 250 mg Amber Glass Bottle
CC33958DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC33958CB Supplier Thermo Scientific Chemicals Supplier No. CC33958CB
Add to Cart

Chemical Identifiers

CAS 857283-76-2
Molecular Formula C15H22N2
Molecular Weight (g/mol) 230.36
MDL Number MFCD07772814
InChI Key UNFZQBXRXGLJLF-UHFFFAOYSA-N
Synonym n-4-piperidin-1-ylbenzyl cyclopropanamine, n-4-piperidin-1-yl phenyl methyl cyclopropanamine, cyclopropyl 4-piperidylphenyl methyl amine, n-4-piperidin-1-yl benzyl cyclopropanamine, benzenemethanamine,n-cyclopropyl-4-1-piperidinyl, n-4-piperidin-1-ylphenyl methyl cyclopropanamine
PubChem CID 7162339
IUPAC Name N-{[4-(piperidin-1-yl)phenyl]methyl}cyclopropanamine
SMILES C(NC1CC1)C1=CC=C(C=C1)N1CCCCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 230.35
Percent Purity 95%
Chemical Name or Material N-(4-piperidin-1-ylbenzyl)cyclopropanamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.