Learn More
Cayman Chemical N-(4-acetamidophenyl)-Indomethacin amide, Purity: ≥98%, CAS: 261766-23-8, 50 mg
SDP
Catalog No. 502894128
Encompass_Preferred
N-4AIA is one of several aromatic amides of indomethacin reported to be potent and selective reversible inhibitors of COX-2.{8243} N-4AIA inhibits human recombinant and ovine COX-2 with IC50 values of 0.1 and 0.625 µM, respectively.{8243,8513} It is about 80 times less potent as an inhibitor of ovine COX-1 than ovine COX-2.
Catalog No. 50-289-4128 Supplier Cayman Chemical Supplier No. 7027850 MG
N-4AIA is one of several aromatic amides of indomethacin reported to be potent and selective reversible inhibitors of COX-2.{8243} N-4AIA inhibits human recombinant and ovine COX-2 with IC50 values of 0.1 and 0.625 µM, respectively.{8243,8513} It is about 80 times less potent as an inhibitor of ovine COX-1 than ovine COX-2.