Learn More
Cayman Chemical N 4AmInophenylmethyladen 10mg
SDP
Catalog No. 502941462
Encompass_Preferred
An adenosine derivative; has been used as an intermediate in the synthesis of [125I]-N6-4-aminobenzyladenosine
Catalog No. 50-294-1462 Supplier Cayman Chemical Supplier No. 2841710 MG
An adenosine derivative; has been used as an intermediate in the synthesis of [125I]-N6-4-aminobenzyladenosine