Learn More
Medchemexpress LLC N-[5-({2-[(cyclopropylcarbonyl)amino]-imidazo[1,2-b]pyridazin-6-yl}oxy)-2-methylphenyl]-1,3-dimethyl-1H-pyrazole-5-... | 1005780-62-0 | MFCD23160045 | 99.7% | 445.47 g·mol⁻1 | C23H23N7O3 | 10 MG
SDP
Catalog No. 5000287464
Encompass_Preferred
TAK-593 is a small-molecule tyrosine kinase inhibitor originally developed by Takeda Pharmaceutical Company. It potently inhibits vascular endothelial growth factor receptors and platelet-derived growth factor receptors and has demonstrated anti-angiogenic and antitumor activity in preclinical studies. Reported biochemical IC50 values include 3.2 nM (VEGFR1), 0.95 nM (VEGFR2), 1.1 nM (VEGFR3), 4.3 nM (PDGFRα), and 13 nM (PDGFRβ).
  • Potent VEGFR and PDGFR inhibition (VEGFR2 IC50 ≈ 0.95 nM).
  • High biochemical selectivity toward receptor tyrosine kinases.
  • Reported molecular formula C23H23N7O3 and molecular weight ≈ 445.47 g·mol⁻1.
  • High purity, typically reported as 99.66% by supplier.
  • Provided with analytical documentation such as LC-MS.
  • Suitable for preclinical studies of angiogenesis and tumor biology.
Catalog No. 50-002-87464 Supplier Medchemexpress LLC Supplier No. HY1550610MG
TAK-593 is a small-molecule tyrosine kinase inhibitor originally developed by Takeda Pharmaceutical Company. It potently inhibits vascular endothelial growth factor receptors and platelet-derived growth factor receptors and has demonstrated anti-angiogenic and antitumor activity in preclinical studies. Reported biochemical IC50 values include 3.2 nM (VEGFR1), 0.95 nM (VEGFR2), 1.1 nM (VEGFR3), 4.3 nM (PDGFRα), and 13 nM (PDGFRβ).
  • Potent VEGFR and PDGFR inhibition (VEGFR2 IC50 ≈ 0.95 nM).
  • High biochemical selectivity toward receptor tyrosine kinases.
  • Reported molecular formula C23H23N7O3 and molecular weight ≈ 445.47 g·mol⁻1.
  • High purity, typically reported as 99.66% by supplier.
  • Provided with analytical documentation such as LC-MS.
  • Suitable for preclinical studies of angiogenesis and tumor biology.