Learn More
Medchemexpress LLC N-{5-[3-(3-hydroxypiperidin-1-yl)-1,2,4-oxadiazol-5-yl]-4-methyl-1,3-thiazol-2-yl}acetamide | 1292300-75-4 | 99.0% | 10 MG
SDP
Catalog No. 5000231940
Encompass_Preferred
TASP0415914 is a small-molecule PI3Kγ inhibitor supplied as a powder for research use. It shows potent PI3Kγ activity (IC50 29 nM) and additional Akt inhibition (IC50 294 nM). The compound has molecular formula C13H17N5O3S, molecular weight 323.37 g/mol, and is typically provided at high purity for laboratory studies.
  • Potent PI3Kγ inhibitor with reported IC50 of 29 nM.
  • Also inhibits Akt with reported IC50 of 294 nM.
  • Molecular formula C13H17N5O3S and molecular weight 323.37 g/mol.
  • High purity, about 99.0% by HPLC.
  • Supplied as a powder in a 10 MG unit for laboratory research.
  • Store frozen for long-term stability; short-term storage at 4°C.
Catalog No. 50-002-31940 Supplier Medchemexpress LLC Supplier No. HY12043810MG
TASP0415914 is a small-molecule PI3Kγ inhibitor supplied as a powder for research use. It shows potent PI3Kγ activity (IC50 29 nM) and additional Akt inhibition (IC50 294 nM). The compound has molecular formula C13H17N5O3S, molecular weight 323.37 g/mol, and is typically provided at high purity for laboratory studies.
  • Potent PI3Kγ inhibitor with reported IC50 of 29 nM.
  • Also inhibits Akt with reported IC50 of 294 nM.
  • Molecular formula C13H17N5O3S and molecular weight 323.37 g/mol.
  • High purity, about 99.0% by HPLC.
  • Supplied as a powder in a 10 MG unit for laboratory research.
  • Store frozen for long-term stability; short-term storage at 4°C.