Learn More
Medchemexpress LLC N-{5-[3-(3-hydroxypiperidin-1-yl)-1,2,4-oxadiazol-5-yl]-4-methyl-1,3-thiazol-2-yl}acetamide | 1292300-75-4 | 99.0% | 25 MG
SDP
Catalog No. 5000234518
Encompass_Preferred
TASP0415914 is a small-molecule PI3Kγ inhibitor supplied as a white to off-white solid for research use. The compound has molecular formula C13H17N5O3S, molecular weight 323.37 g/mol, and a reported purity of 99.0%.
  • PI3Kγ inhibitor for biochemical and cellular research.
  • White to off-white solid, stable at room temperature.
  • Molecular weight 323.37 g/mol; formula C13H17N5O3S.
  • Reported purity 99.0% as supplied.
  • Available as a 25 MG quantity for laboratory studies.
Catalog No. 50-002-34518 Supplier Medchemexpress LLC Supplier No. HY12043825MG
TASP0415914 is a small-molecule PI3Kγ inhibitor supplied as a white to off-white solid for research use. The compound has molecular formula C13H17N5O3S, molecular weight 323.37 g/mol, and a reported purity of 99.0%.
  • PI3Kγ inhibitor for biochemical and cellular research.
  • White to off-white solid, stable at room temperature.
  • Molecular weight 323.37 g/mol; formula C13H17N5O3S.
  • Reported purity 99.0% as supplied.
  • Available as a 25 MG quantity for laboratory studies.