Learn More
Medchemexpress LLC N-{5-[3-(3-hydroxypiperidin-1-yl)-1,2,4-oxadiazol-5-yl]-4-methyl-1,3-thiazol-2-yl}acetamide | 1292300-75-4 | 99.0% | 5 MG
SDP
Catalog No. 5000234353
Encompass_Preferred
TASP0415914 is a research-grade small-molecule inhibitor primarily targeting PI3Kγ, with additional activity against Akt. It is supplied as a high-purity solid suitable for biochemical and cellular studies and is typically packaged in 5 mg quantities.
  • Potent PI3Kγ inhibition (IC50 = 29 nM).
  • Additional Akt inhibitory activity (IC50 = 294 nM).
  • High purity suitable for research applications.
  • Powder stable at -20°C for long-term storage; in-solution storage at -80°C recommended.
  • Molecular weight 323.37 g/mol; formula C13H17N5O3S.
Catalog No. 50-002-34353 Supplier Medchemexpress LLC Supplier No. HY1204385MG
TASP0415914 is a research-grade small-molecule inhibitor primarily targeting PI3Kγ, with additional activity against Akt. It is supplied as a high-purity solid suitable for biochemical and cellular studies and is typically packaged in 5 mg quantities.
  • Potent PI3Kγ inhibition (IC50 = 29 nM).
  • Additional Akt inhibitory activity (IC50 = 294 nM).
  • High purity suitable for research applications.
  • Powder stable at -20°C for long-term storage; in-solution storage at -80°C recommended.
  • Molecular weight 323.37 g/mol; formula C13H17N5O3S.