Learn More
Medchemexpress LLC N-{5-[3-(3-hydroxypiperidin-1-yl)-1,2,4-oxadiazol-5-yl]-4-methyl-1,3-thiazol-2-yl}acetamide | 1292300-75-4 | 99.0% | 50 MG
SDP
Catalog No. 5000234451
Encompass_Preferred
TASP0415914 is a small-molecule research inhibitor of PI3Kγ with reported oral activity and secondary Akt inhibition. It is supplied as a high-purity solid for preclinical studies of PI3K/Akt signaling and inflammatory disease models.
  • Potent PI3Kγ inhibitor with IC50 29 nM and Akt activity IC50 294 nM.
  • High purity of 99.0% suitable for research use.
  • White to off-white solid, molecular weight 323.37 g/mol, formula C13H17N5O3S.
  • Storage recommendations: powder at -20°C for long-term storage; in solvent at -80°C for best stability.
  • Intended for preclinical and mechanistic studies; not for human use.
Catalog No. 50-002-34451 Supplier Medchemexpress LLC Supplier No. HY12043850MG
TASP0415914 is a small-molecule research inhibitor of PI3Kγ with reported oral activity and secondary Akt inhibition. It is supplied as a high-purity solid for preclinical studies of PI3K/Akt signaling and inflammatory disease models.
  • Potent PI3Kγ inhibitor with IC50 29 nM and Akt activity IC50 294 nM.
  • High purity of 99.0% suitable for research use.
  • White to off-white solid, molecular weight 323.37 g/mol, formula C13H17N5O3S.
  • Storage recommendations: powder at -20°C for long-term storage; in solvent at -80°C for best stability.
  • Intended for preclinical and mechanistic studies; not for human use.