Learn More
Medchemexpress LLC N-[5-(3,5-dicyano-1,4-dihydro-1,2,6-trimethyl-4-pyridinyl)-6-fluoro-7-methyl-1H-indazol-3-yl] | 2377576-35-5 | 99.7% | 468.53 | C27H25FN6O | 5 MG
SDP
Catalog No. 5000264722
Encompass_Preferred
BAY-3827 is a potent, selective small-molecule inhibitor of AMP-activated protein kinase (AMPK) used as a chemical probe in preclinical research. It demonstrates antiproliferative activity in androgen-dependent prostate cancer cell models and is supplied as a solid with reported high purity for biochemical and cellular studies.
  • Potent and selective AMPK inhibitor.
  • Demonstrated antiproliferative activity in androgen-dependent prostate cancer cell lines.
  • Suitable for biochemical and cellular assays as a research chemical.
  • Provided as a solid in small-milligram quantities for early-stage experiments.
  • Reported high purity suitable for analytical and biological studies.
Catalog No. 50-002-64722 Supplier Medchemexpress LLC Supplier No. HY1120835MG
BAY-3827 is a potent, selective small-molecule inhibitor of AMP-activated protein kinase (AMPK) used as a chemical probe in preclinical research. It demonstrates antiproliferative activity in androgen-dependent prostate cancer cell models and is supplied as a solid with reported high purity for biochemical and cellular studies.
  • Potent and selective AMPK inhibitor.
  • Demonstrated antiproliferative activity in androgen-dependent prostate cancer cell lines.
  • Suitable for biochemical and cellular assays as a research chemical.
  • Provided as a solid in small-milligram quantities for early-stage experiments.
  • Reported high purity suitable for analytical and biological studies.