Learn More
Medchemexpress LLC N-[5-[[3-cyano-7-(cyclopropylamino)pyrazolo[1,5-a]pyrimidin-5-yl]amino]-2-methylphenyl]propanamide | 2470424-39-4 | 99.7% | C20H21N7O | 10MG
SDP
Catalog No. 5000259288
Encompass_Preferred
SGC-CK2-1 is a selective, cell-active ATP-competitive chemical probe for casein kinase 2 (CK2) used in research to investigate CK2 signaling and neurodegenerative disease-related pathways. It shows low-nanomolar potency in cellular assays and is suitable for mechanistic and cell-based studies.
  • Low-nanomolar cellular potency against CK2 isoforms.
  • High selectivity for CK2 over other kinases.
  • Cell-permeable and active in cell-based assays.
  • Suitable for mechanistic studies of CK2 signaling and neurodegeneration.
  • Stable solid form with recommended cold storage for long-term retention.
Catalog No. 50-002-59288 Supplier Medchemexpress LLC Supplier No. HY13900410MG
SGC-CK2-1 is a selective, cell-active ATP-competitive chemical probe for casein kinase 2 (CK2) used in research to investigate CK2 signaling and neurodegenerative disease-related pathways. It shows low-nanomolar potency in cellular assays and is suitable for mechanistic and cell-based studies.
  • Low-nanomolar cellular potency against CK2 isoforms.
  • High selectivity for CK2 over other kinases.
  • Cell-permeable and active in cell-based assays.
  • Suitable for mechanistic studies of CK2 signaling and neurodegeneration.
  • Stable solid form with recommended cold storage for long-term retention.