Learn More
Medchemexpress LLC N-[5-({[(3-fluorophenyl)carbamoyl]amino}methyl)-2-methylphenyl]imidazo[1,2-a]pyridine-3-carboxamide | 1644069-80-6 | MFCD31567545 | 99.0% | 417.44 | C23H20FN5O2 | 1 ML
SDP
Catalog No. 5000300836
Encompass_Preferred
DDR Inhibitor is a small-molecule discoidin domain receptor (DDR) inhibitor for research applications. It potently inhibits DDR2 (IC50 3.3 nM), shows partial activity against DDR1 (53% inhibition at 1.5 nM), and inhibits collagen I-induced DDR2 phosphorylation in cells (EC50 8.7 nM). Available as concentrated DMSO solution or as solid for biochemical and cell-based studies.
  • Potent DDR2 inhibition (IC50 3.3 nM).
  • Partial DDR1 activity (53% inhibition at 1.5 nM).
  • Cell-active inhibition of collagen I-induced DDR2 phosphorylation (EC50 8.7 nM).
  • Available as 10 mM solution in DMSO or as solid quantities.
  • High reported purity (>99% by HPLC) with COA available.
  • Suitable for biochemical and cell-based DDR signaling studies.
Catalog No. 50-003-00836 Supplier Medchemexpress LLC Supplier No. HYW01893110MM/1ML
DDR Inhibitor is a small-molecule discoidin domain receptor (DDR) inhibitor for research applications. It potently inhibits DDR2 (IC50 3.3 nM), shows partial activity against DDR1 (53% inhibition at 1.5 nM), and inhibits collagen I-induced DDR2 phosphorylation in cells (EC50 8.7 nM). Available as concentrated DMSO solution or as solid for biochemical and cell-based studies.
  • Potent DDR2 inhibition (IC50 3.3 nM).
  • Partial DDR1 activity (53% inhibition at 1.5 nM).
  • Cell-active inhibition of collagen I-induced DDR2 phosphorylation (EC50 8.7 nM).
  • Available as 10 mM solution in DMSO or as solid quantities.
  • High reported purity (>99% by HPLC) with COA available.
  • Suitable for biochemical and cell-based DDR signaling studies.