Learn More
Medchemexpress LLC N-[5-[4-chloro-3-[(2-hydroxyethyl)sulfamoyl]phenyl]-4-methylthiazol-2-yl]acetamide | 593960-11-3 | >98.0% | 10 MM 1 ML
SDP
Catalog No. 5000234139
Encompass_Preferred
PIK-93 is a potent, synthetic inhibitor of phosphatidylinositol 4-kinase IIIβ (PI4KIIIβ) with nanomolar activity against PI4K and PI3K isoforms. Reported IC50 values are 19 nM (PI4KIIIβ), 16 nM (PI3Kγ), and 39 nM (PI3Kα). Molecular formula C14H16ClN3O4S2; molecular weight 389.88 g/mol. Typically supplied as a 10 mM solution in DMSO (1 mL) for in vitro use.
  • Potent PI4KIIIβ inhibitor with nanomolar potency.
  • Inhibits PI3Kγ and PI3Kα at low nanomolar concentrations.
  • Supplied as a 10 mM DMSO solution (1 mL) for in vitro assays.
  • High purity suitable for biochemical and cell-based studies (supplier-dependent).
Catalog No. 50-002-34139 Supplier Medchemexpress LLC Supplier No. HY1204610MM 1ML
Add to Cart
Add to Cart
PIK-93 is a potent, synthetic inhibitor of phosphatidylinositol 4-kinase IIIβ (PI4KIIIβ) with nanomolar activity against PI4K and PI3K isoforms. Reported IC50 values are 19 nM (PI4KIIIβ), 16 nM (PI3Kγ), and 39 nM (PI3Kα). Molecular formula C14H16ClN3O4S2; molecular weight 389.88 g/mol. Typically supplied as a 10 mM solution in DMSO (1 mL) for in vitro use.
  • Potent PI4KIIIβ inhibitor with nanomolar potency.
  • Inhibits PI3Kγ and PI3Kα at low nanomolar concentrations.
  • Supplied as a 10 mM DMSO solution (1 mL) for in vitro assays.
  • High purity suitable for biochemical and cell-based studies (supplier-dependent).