Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[5-[4-chloro-3-(2-hydroxyethylsulfamoyl)phenyl]-4-methyl-1,3-thiazol-2-yl]acetamide | 593960-11-3 | >98.0% | 100 MG
SDP

Catalog No. 5000231848
Encompass_Preferred

PIK-93 is a small-molecule kinase inhibitor used in research to inhibit PI4KIIIβ and certain PI3K isoforms. It is reported to inhibit PI4KIIIβ with an IC50 of 19 nM, PI3Kγ with an IC50 of 16 nM, and PI3Kα with an IC50 of 39 nM.

  • Potent PI4KIIIβ inhibitor; IC50 = 19 nM.
  • Also inhibits PI3Kγ (IC50 = 16 nM) and PI3Kα (IC50 = 39 nM).
  • Chemical formula C14H16ClN3O4S2; molecular weight 389.9 g/mol.
  • Typically supplied as a solid for research applications.
  • Purity commonly reported ≥98% by suppliers.
  • Useful for studies of PI4K/PI3K signaling and viral replication pathways.

Catalog No. 50-002-31848 Supplier Medchemexpress LLC Supplier No. HY12046100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PIK-93 is a small-molecule kinase inhibitor used in research to inhibit PI4KIIIβ and certain PI3K isoforms. It is reported to inhibit PI4KIIIβ with an IC50 of 19 nM, PI3Kγ with an IC50 of 16 nM, and PI3Kα with an IC50 of 39 nM.

  • Potent PI4KIIIβ inhibitor; IC50 = 19 nM.
  • Also inhibits PI3Kγ (IC50 = 16 nM) and PI3Kα (IC50 = 39 nM).
  • Chemical formula C14H16ClN3O4S2; molecular weight 389.9 g/mol.
  • Typically supplied as a solid for research applications.
  • Purity commonly reported ≥98% by suppliers.
  • Useful for studies of PI4K/PI3K signaling and viral replication pathways.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.