Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[5-[4-chloro-3-(2-hydroxyethylsulfamoyl)phenyl]-4-methyl-1,3-thiazol-2-yl]acetamide | 593960-11-3 | 99.8% | 25 MG
SDP

Catalog No. 5000231849
Encompass_Preferred

PIK-93 is a small-molecule kinase inhibitor that potently inhibits PI4KIIIβ and also shows activity against PI3K isoforms. It is supplied for research use to probe PI4K/PI3K-related signaling pathways in biochemical and cell-based assays.

  • Molecular formula: C14H16ClN3O4S2
  • Molecular weight: 389.9 g/mol
  • CAS number: 593960-11-3
  • Purity: approximately 99.8% (reported by supplier)
  • Appearance: white to off-white solid
  • Storage: powder -20°C for long-term storage; in solvent -80°C recommended
  • Potency: PI4KIIIβ IC50 ≈ 19 nM; PI3Kγ IC50 ≈ 16 nM; PI3Kα IC50 ≈ 39 nM
  • Typical use: biochemical kinase assays and cell signaling studies (research use only)

Catalog No. 50-002-31849 Supplier Medchemexpress LLC Supplier No. HY1204625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PIK-93 is a small-molecule kinase inhibitor that potently inhibits PI4KIIIβ and also shows activity against PI3K isoforms. It is supplied for research use to probe PI4K/PI3K-related signaling pathways in biochemical and cell-based assays.

  • Molecular formula: C14H16ClN3O4S2
  • Molecular weight: 389.9 g/mol
  • CAS number: 593960-11-3
  • Purity: approximately 99.8% (reported by supplier)
  • Appearance: white to off-white solid
  • Storage: powder -20°C for long-term storage; in solvent -80°C recommended
  • Potency: PI4KIIIβ IC50 ≈ 19 nM; PI3Kγ IC50 ≈ 16 nM; PI3Kα IC50 ≈ 39 nM
  • Typical use: biochemical kinase assays and cell signaling studies (research use only)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.