Learn More
Medchemexpress LLC N-[5-[4-chloro-3-(2-hydroxyethylsulfamoyl)phenyl]-4-methyl-1,3-thiazol-2-yl]acetamide | 593960-11-3 | 99.8% | 5 MG
SDP
Catalog No. 5000233364
Encompass_Preferred
PIK-93 is a small-molecule kinase inhibitor used in research to probe phosphatidylinositol 4-kinase (PI4KIIIβ) and PI3K signaling pathways. It displays low-nanomolar potency and is supplied as a solid for laboratory use.
  • Potent PI4KIIIβ inhibitor (IC50 ≈ 19 nM).
  • Also inhibits PI3Kγ and PI3Kα (IC50 ≈ 16 nM and ≈ 39 nM).
  • Molecular formula C14H16ClN3O4S2; molecular weight ≈ 389.9 g/mol.
  • High reported purity (vendor-reported) and stability when stored frozen.
  • Intended for research use; follow standard laboratory safety and handling procedures.
Catalog No. 50-002-33364 Supplier Medchemexpress LLC Supplier No. HY120465MG
PIK-93 is a small-molecule kinase inhibitor used in research to probe phosphatidylinositol 4-kinase (PI4KIIIβ) and PI3K signaling pathways. It displays low-nanomolar potency and is supplied as a solid for laboratory use.
  • Potent PI4KIIIβ inhibitor (IC50 ≈ 19 nM).
  • Also inhibits PI3Kγ and PI3Kα (IC50 ≈ 16 nM and ≈ 39 nM).
  • Molecular formula C14H16ClN3O4S2; molecular weight ≈ 389.9 g/mol.
  • High reported purity (vendor-reported) and stability when stored frozen.
  • Intended for research use; follow standard laboratory safety and handling procedures.