Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(5-(((4-Me-1,2,5-thiadiazol-3-yl)thio)methyl)-1,3,4-thiadiazol-2-yl)-4-(4-(CF3)Ph)thiazol-2-amine | 2809469-05-2 | 99.0% | 501.56 g·mol⁻¹ | C21H18F3NO4S3 | 100 MG
SDP

Catalog No. 5000245643
Encompass_Preferred

NAZ2329 is a research-grade, cell-permeable small-molecule inhibitor that allosterically and preferentially targets the receptor-type protein tyrosine phosphatases PTPRZ and PTPRG. It has demonstrated inhibition of glioblastoma cell growth and suppression of stem cell-like properties in published studies. Supplied as a purified powder for laboratory research use.

  • Allosteric inhibitor of PTPRZ and PTPRG.
  • Demonstrated antiproliferative activity in glioblastoma models.
  • High reported purity (99.02%).
  • Soluble in DMSO (100 mg/mL); in vivo formulation protocols available.
  • Recommended storage: powder at -20 °C for long-term stability.

Catalog No. 50-002-45643 Supplier Medchemexpress LLC Supplier No. HY103693100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NAZ2329 is a research-grade, cell-permeable small-molecule inhibitor that allosterically and preferentially targets the receptor-type protein tyrosine phosphatases PTPRZ and PTPRG. It has demonstrated inhibition of glioblastoma cell growth and suppression of stem cell-like properties in published studies. Supplied as a purified powder for laboratory research use.

  • Allosteric inhibitor of PTPRZ and PTPRG.
  • Demonstrated antiproliferative activity in glioblastoma models.
  • High reported purity (99.02%).
  • Soluble in DMSO (100 mg/mL); in vivo formulation protocols available.
  • Recommended storage: powder at -20 °C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.