Learn More
Medchemexpress LLC N-5-benzothiazolyl-6-[(1-methylethyl)sulfonyl]-4-quinolinamine | 1346546-69-7 | MFCD30481302 | 99.9% | 383.49 g/mol | C19H17N3O2S2 | 1 MG
SDP
Catalog No. 5000248033
Encompass_Preferred
A potent, selective small-molecule inhibitor of receptor-interacting protein kinase 3 (RIPK3) for research use to block necroptosis and study RIPK3-dependent signaling. The compound has molecular weight 383.49 g/mol, chemical formula C19H17N3O2S2, CAS 1346546-69-7, and is supplied at high purity, making it suitable for biochemical and cell-based assays.
  • Selective RIPK3 kinase inhibitor with low-nanomolar potency.
  • Potent biochemical inhibition (IC50 ≈ 1.8 nM).
  • High purity (99.9%) suitable for sensitive assays.
  • Available in small research quantities, including 1 mg units.
  • Stable under recommended storage conditions for powder and solutions.
Catalog No. 50-002-48033 Supplier Medchemexpress LLC Supplier No. HY1018721MG
A potent, selective small-molecule inhibitor of receptor-interacting protein kinase 3 (RIPK3) for research use to block necroptosis and study RIPK3-dependent signaling. The compound has molecular weight 383.49 g/mol, chemical formula C19H17N3O2S2, CAS 1346546-69-7, and is supplied at high purity, making it suitable for biochemical and cell-based assays.
  • Selective RIPK3 kinase inhibitor with low-nanomolar potency.
  • Potent biochemical inhibition (IC50 ≈ 1.8 nM).
  • High purity (99.9%) suitable for sensitive assays.
  • Available in small research quantities, including 1 mg units.
  • Stable under recommended storage conditions for powder and solutions.