Learn More
Medchemexpress LLC N-[(5-bromo-2-methoxyphenyl)methyl]adamantan-1-amine | 332108-65-3 | 98.5% | 350.29 g/mol | C18H24BrNO | 50MG
SDP
Catalog No. 5000237140
Encompass_Preferred
ABMA (N-[(5-bromo-2-methoxyphenyl)methyl]adamantan-1-amine) is a small-molecule inhibitor that protects cells against a range of intracellular toxins and pathogens. It is supplied for laboratory research as a solid and as standardized DMSO solutions, with pack sizes suitable for bench-scale experiments.
Catalog No. 50-002-37140 Supplier Medchemexpress LLC Supplier No. HY12480150MG
ABMA (N-[(5-bromo-2-methoxyphenyl)methyl]adamantan-1-amine) is a small-molecule inhibitor that protects cells against a range of intracellular toxins and pathogens. It is supplied for laboratory research as a solid and as standardized DMSO solutions, with pack sizes suitable for bench-scale experiments.