Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(5-chloropyridin-2-yl)-2-(4-(N,N-dimethylcarbamimidoyl)benzamido)-5-methoxybenzamide | 330942-05-7 | MFCD16038040 | >=98.0% | 451.91 | C23H22ClN5O3 | 1 ML
SDP

Catalog No. 5000245756
Encompass_Preferred

Betrixaban is a potent, selective factor Xa inhibitor used as a research reagent for anticoagulation studies. The compound is characterized by a reported IC50 of 1.5 nM against factor Xa and a molecular weight of 451.91; it is typically handled as a research-grade small molecule and dissolved in DMSO for stock preparations.

  • Potent factor Xa inhibitor with reported IC50 of 1.5 nM.
  • Research-grade small molecule for in vitro and in vivo studies.
  • Molecular formula C23H22ClN5O3 and molecular weight 451.91.
  • Recommended solvent for stocks: DMSO; example preparations provided for 10 mM solutions.
  • Storage: powder at -20 °C (long term) or 4 °C (short term); in solvent at -80 °C or -20 °C depending on duration.

Catalog No. 50-002-45756 Supplier Medchemexpress LLC Supplier No. HY1026810MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Betrixaban is a potent, selective factor Xa inhibitor used as a research reagent for anticoagulation studies. The compound is characterized by a reported IC50 of 1.5 nM against factor Xa and a molecular weight of 451.91; it is typically handled as a research-grade small molecule and dissolved in DMSO for stock preparations.

  • Potent factor Xa inhibitor with reported IC50 of 1.5 nM.
  • Research-grade small molecule for in vitro and in vivo studies.
  • Molecular formula C23H22ClN5O3 and molecular weight 451.91.
  • Recommended solvent for stocks: DMSO; example preparations provided for 10 mM solutions.
  • Storage: powder at -20 °C (long term) or 4 °C (short term); in solvent at -80 °C or -20 °C depending on duration.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.