Learn More
AdipoGen N-(5-Fluoresceinyl)-maleinimide, 75350-46-8, MFCD00077345, 50 mg
SDP
Catalog No. 50596506
Encompass_Preferred
MF: C24H13NO7, MW: 427.36, ≥90%, Appearance: Orange to brown solid. Green fluorescent thiol-reactive dye (Ex/Em: 494/519nm) for bioconjugation, widely used as protein and peptide fluorescent labeling reagent, easily conjugating fluorescein. The reaction requires very mild condition and is highly specific. The maleimide group reacts optimally with sulfhydryl groups on cysteine side chains at pH 7, forming a stable thioether bond. The compound can be used to label proteins for the detection of conformational changes, assembly of multisubunit complexes, and ligand-binding processes.
Catalog No. 50-596-506 Supplier AdipoGen Supplier No. CDXF0004M050
MF: C24H13NO7, MW: 427.36, ≥90%, Appearance: Orange to brown solid. Green fluorescent thiol-reactive dye (Ex/Em: 494/519nm) for bioconjugation, widely used as protein and peptide fluorescent labeling reagent, easily conjugating fluorescein. The reaction requires very mild condition and is highly specific. The maleimide group reacts optimally with sulfhydryl groups on cysteine side chains at pH 7, forming a stable thioether bond. The compound can be used to label proteins for the detection of conformational changes, assembly of multisubunit complexes, and ligand-binding processes.