Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-((5-fluoro-2,3-dihydro-1-benzofuran-4-yl)methyl)-8-(2-methylpyridin-3-yl)triazolopyrimidin-5-amine | 1951408-58-4 | MFCD31630846 | 99.2% | 376.39 g/mol | C20H17FN6O | 100 MG
SDP

Catalog No. 5000245168
Encompass_Preferred

MAK683 is a small-molecule embryonic ectoderm development (EED) inhibitor that binds the EED subunit of PRC2 and acts as an allosteric antagonist of PRC2 function. Supplied for research use, it supports biochemical and cellular studies of epigenetic regulation.

  • Allosteric EED/PRC2 inhibitor for epigenetic research.
  • Reported purity: 99.18% (manufacturer data).
  • Chemical formula: C20H17FN6O; molecular weight: 376.39 g/mol.
  • Soluble in DMSO; typical handling for small-molecule reagents.
  • Available in multiple pack sizes for laboratory use.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-45168 Supplier Medchemexpress LLC Supplier No. HY103663100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MAK683 is a small-molecule embryonic ectoderm development (EED) inhibitor that binds the EED subunit of PRC2 and acts as an allosteric antagonist of PRC2 function. Supplied for research use, it supports biochemical and cellular studies of epigenetic regulation.

  • Allosteric EED/PRC2 inhibitor for epigenetic research.
  • Reported purity: 99.18% (manufacturer data).
  • Chemical formula: C20H17FN6O; molecular weight: 376.39 g/mol.
  • Soluble in DMSO; typical handling for small-molecule reagents.
  • Available in multiple pack sizes for laboratory use.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.