Learn More
Medchemexpress LLC N-(5-Sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide-2,2,2-d3 | 1189904-01-5 | 99.9% | 5 MG
SDP
Catalog No. 5000407730
Encompass_Preferred
Acetazolamide-d3 is a deuterium-labeled version of Acetazolamide, a potent carbonic anhydrase (CA) IX inhibitor with an IC50 of 30 nM for hCA IX. It exhibits diuretic, antihypertensive, and anti-gonococcal activities. This product is intended for research use only and has not been fully validated for medical applications.
  • Deuterium labeled Acetazolamide
  • Potent carbonic anhydrase (CA) IX inhibitor
  • IC50 of 30 nM for hCA IX
  • Exhibits diuretic activity
  • Exhibits antihypertensive activity
  • Exhibits anti-gonococcal activity
  • Suitable for research applications
Catalog No. 50-004-07730 Supplier Medchemexpress LLC Supplier No. HYB0782S5MG
Acetazolamide-d3 is a deuterium-labeled version of Acetazolamide, a potent carbonic anhydrase (CA) IX inhibitor with an IC50 of 30 nM for hCA IX. It exhibits diuretic, antihypertensive, and anti-gonococcal activities. This product is intended for research use only and has not been fully validated for medical applications.
  • Deuterium labeled Acetazolamide
  • Potent carbonic anhydrase (CA) IX inhibitor
  • IC50 of 30 nM for hCA IX
  • Exhibits diuretic activity
  • Exhibits antihypertensive activity
  • Exhibits anti-gonococcal activity
  • Suitable for research applications