Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[6-(4-chlorophenoxy)hexyl]-N'-cyano-N''-4-pyridinyl-guanidine | 200484-11-3 | MFCD03700766 | 99.3% | 371.86 g/mol | C19H22ClN5O | 10 MG
SDP

Catalog No. 5000212955
Encompass_Preferred

CHS-828 (GMX1778) is a small-molecule competitive inhibitor of nicotinamide phosphoribosyltransferase (NAMPT) supplied for research use. It is provided as a high-purity solid and in solution formats for biochemical and cellular studies, with documented storage and stability recommendations for powder and solvent preparations.

  • Potent NAMPT inhibitor with low-nanomolar activity.
  • High purity (>99.2%).
  • Molecular weight 371.86 g/mol.
  • Molecular formula C19H22ClN5O.
  • Available as a solid powder and as a solution for convenience.
  • Storage conditions support long-term stability at low temperatures.
  • Intended for research and analytical applications only.

Catalog No. 50-002-12955 Supplier Medchemexpress LLC Supplier No. HY1007910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHS-828 (GMX1778) is a small-molecule competitive inhibitor of nicotinamide phosphoribosyltransferase (NAMPT) supplied for research use. It is provided as a high-purity solid and in solution formats for biochemical and cellular studies, with documented storage and stability recommendations for powder and solvent preparations.

  • Potent NAMPT inhibitor with low-nanomolar activity.
  • High purity (>99.2%).
  • Molecular weight 371.86 g/mol.
  • Molecular formula C19H22ClN5O.
  • Available as a solid powder and as a solution for convenience.
  • Storage conditions support long-term stability at low temperatures.
  • Intended for research and analytical applications only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.