Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(6-Aminohexyl)-1-naphthalenesulfonamide Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A241025MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A241025MG 25 mg
A2410500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A241025MG Supplier TCI America Supplier No. A241025MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 61714-25-8
Molecular Formula C16H23ClN2O2S
Molecular Weight (g/mol) 342.88
MDL Number MFCD00058043
InChI Key HOCSVIGHWPLMFC-UHFFFAOYSA-N
Synonym w-5 hydrochloride, n-6-aminohexyl-1-naphthalenesulfonamide hydrochloride, w-5, hydrochloride, n-6-aminohexyl naphthalene-1-sulfonamide hydrochloride, n-6-aminohexyl-1-naphthalenesulphonamide hydrochloride, n-6-aminohexyl-1-naphthalenesulfonamide hcl, n-6-aminohexyl-1-naphthalenesulfonamide, hcl, acmc-209mvm, n-6-aminohexyl-1-naphthalenesulfonamide monohydrochlorde, 1-naphthalenesulfonamide, n-6-aminohexyl-, monohydrochlorde
PubChem CID 173829
IUPAC Name hydrogen N-(6-aminohexyl)naphthalene-1-sulfonamide chloride
SMILES [H+].[Cl-].NCCCCCCNS(=O)(=O)C1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 164°C
Color White-Yellow
Quantity 25 mg
Formula Weight 342.88
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(6-Aminohexyl)-1-naphthalenesulfonamide Hydrochloride
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.