Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(6-Aminohexyl)-2-nitrobenzenesulfonamide Hydrochloride 95.0+%, TCI America™
SDP

Catalog No. A16621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A16621G Supplier TCI America Supplier No. A16621G

Chemical Identifiers

CAS 95915-82-5
Molecular Formula C12H20ClN3O4S
Molecular Weight (g/mol) 337.82
MDL Number MFCD06797045
InChI Key CRQMJHQZDIEKGD-UHFFFAOYSA-N
Synonym 1-Amino-6-(2-nitrobenzenesulfonamido)hexane Hydrochloride, N-(2-Nitrobenzenesulfonyl)-1,6-hexanediamine Hydrochloride, N-(o-Ns)-1,6-diaminohexane Hydrochloride
PubChem CID 22719658
IUPAC Name N-(6-aminohexyl)-2-nitrobenzene-1-sulfonamide hydrochloride
SMILES Cl.NCCCCCCNS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 164°C
Quantity 1 g
Formula Weight 337.82
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(6-Aminohexyl)-2-nitrobenzenesulfonamide Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.