Learn More
Medchemexpress LLC N-(6-aminohexyl)-5-chloro-1-naphthalenesulfonamide hydrochloride | 61714-27-0 | MFCD00012559 | >98.0% | 377.33 g/mol | C16H21ClN2O2S · HCl | 5 MG
SDP
Catalog No. 5000213355
Encompass_Preferred
W-7 hydrochloride is the hydrochloride salt of W-7, a selective calmodulin antagonist used in biochemical and pharmacological research to inhibit Ca2+-calmodulin-dependent enzymes and modulate calcium-dependent signaling. It is commonly applied in studies of phosphodiesterase regulation, myosin light chain kinase, ion channel function, apoptosis, and vascular relaxation.
  • Selective calmodulin antagonist that inhibits Ca2+-calmodulin-dependent phosphodiesterase and myosin light chain kinase
  • Modulates ion channel activity and is used in arrhythmia research
  • Available as the hydrochloride salt for improved solubility
  • Typical purity >=98% (HPLC)
  • Molecular weight ≈377.33 g/mol
  • Supplied in small research quantities, e.g., 5 mg
Catalog No. 50-002-13355 Supplier Medchemexpress LLC Supplier No. HY1009125MG
W-7 hydrochloride is the hydrochloride salt of W-7, a selective calmodulin antagonist used in biochemical and pharmacological research to inhibit Ca2+-calmodulin-dependent enzymes and modulate calcium-dependent signaling. It is commonly applied in studies of phosphodiesterase regulation, myosin light chain kinase, ion channel function, apoptosis, and vascular relaxation.
  • Selective calmodulin antagonist that inhibits Ca2+-calmodulin-dependent phosphodiesterase and myosin light chain kinase
  • Modulates ion channel activity and is used in arrhythmia research
  • Available as the hydrochloride salt for improved solubility
  • Typical purity >=98% (HPLC)
  • Molecular weight ≈377.33 g/mol
  • Supplied in small research quantities, e.g., 5 mg