Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(6-aminohexyl)-5-chloro-2-naphthalenesulfonamide hydrochloride | 69762-85-2 | 99.7% | 377.33 g/mol | C16H22Cl2N2O2S | 25 MG
SDP

Catalog No. 5000212596
Encompass_Preferred

W-7 isomer hydrochloride is an isomer of W-7 hydrochloride that acts as a selective calmodulin antagonist for research applications. Supplied as a white to off-white solid with high reported purity, it is provided in small pack sizes suitable for assay development and biochemical studies. Handle and store according to recommended conditions and prepare stock solutions following solvent and sterile filtration guidance.

  • High purity (99.65%).
  • Chemical formula C16H22Cl2N2O2S; molecular weight 377.33 g/mol.
  • Appearance: white to off-white solid.
  • Recommended pack sizes include 1 mg to 500 mg, including 25 mg.
  • Storage: powder -20°C (3 years) or 4°C (2 years); aliquot stock solutions to avoid freeze-thaw.
  • For research use only; not intended for human use.

Catalog No. 50-002-12596 Supplier Medchemexpress LLC Supplier No. HY10091325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

W-7 isomer hydrochloride is an isomer of W-7 hydrochloride that acts as a selective calmodulin antagonist for research applications. Supplied as a white to off-white solid with high reported purity, it is provided in small pack sizes suitable for assay development and biochemical studies. Handle and store according to recommended conditions and prepare stock solutions following solvent and sterile filtration guidance.

  • High purity (99.65%).
  • Chemical formula C16H22Cl2N2O2S; molecular weight 377.33 g/mol.
  • Appearance: white to off-white solid.
  • Recommended pack sizes include 1 mg to 500 mg, including 25 mg.
  • Storage: powder -20°C (3 years) or 4°C (2 years); aliquot stock solutions to avoid freeze-thaw.
  • For research use only; not intended for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.