Learn More
Medchemexpress LLC N-(6-aminohexyl)maleimide trifluoroacetate | 731862-92-3 | MFCD11519191 | 99.7% | 310.27 g/mol | C12H17F3N2O4 | 500 MG
SDP
Catalog No. 5000300807
Encompass_Preferred
Mal-C6-amine (TFA) is an alkyl chain-based PROTAC linker supplied as the trifluoroacetate salt for research use. It contains a maleimide reactive terminus and a six-carbon amino linker that enables conjugation to thiols and amines, supporting synthesis of targeted protein degraders and other bioconjugates. The material is provided as a high-purity solid with recommended sealed storage to maintain stability.
  • Alkyl chain linker with maleimide and amino functional groups.
  • Trifluoroacetate salt form for improved handling and stability.
  • High reported purity suitable for synthetic applications.
  • Compatible with common solvents such as DMSO for reaction setup.
  • Intended for research use in PROTAC and bioconjugation workflows.
Catalog No. 50-003-00807 Supplier Medchemexpress LLC Supplier No. HYW018291500MG
Mal-C6-amine (TFA) is an alkyl chain-based PROTAC linker supplied as the trifluoroacetate salt for research use. It contains a maleimide reactive terminus and a six-carbon amino linker that enables conjugation to thiols and amines, supporting synthesis of targeted protein degraders and other bioconjugates. The material is provided as a high-purity solid with recommended sealed storage to maintain stability.
  • Alkyl chain linker with maleimide and amino functional groups.
  • Trifluoroacetate salt form for improved handling and stability.
  • High reported purity suitable for synthetic applications.
  • Compatible with common solvents such as DMSO for reaction setup.
  • Intended for research use in PROTAC and bioconjugation workflows.