Learn More
Medchemexpress LLC N-[6-(biotinamido)hexyl]-3-(2-pyridyldithio)propionamide | 129179-83-5 | MFCD00078541 | 99.7% | C24H37N5O3S3 | 10MG
SDP
Catalog No. 5000259744
Encompass_Preferred
Biotin-HPDP is a thiol-reactive biotinylation reagent that labels free protein thiols through a cleavable disulfide linkage, enabling reversible biotinylation for affinity purification and recovery of native proteins. It is supplied as a white to off-white solid with high reported purity and defined storage recommendations.
  • Thiol-reactive biotinylation reagent for cysteine labeling.
  • Cleavable disulfide linkage enables reversible labeling.
  • Suitable for affinity purification and protein recovery.
  • High reported purity (99.7%).
  • Stable as powder at -20°C for long-term storage.
  • Available in small pack sizes for biochemical assays.
Catalog No. 50-002-59744 Supplier Medchemexpress LLC Supplier No. HY13676910MG
Biotin-HPDP is a thiol-reactive biotinylation reagent that labels free protein thiols through a cleavable disulfide linkage, enabling reversible biotinylation for affinity purification and recovery of native proteins. It is supplied as a white to off-white solid with high reported purity and defined storage recommendations.
  • Thiol-reactive biotinylation reagent for cysteine labeling.
  • Cleavable disulfide linkage enables reversible labeling.
  • Suitable for affinity purification and protein recovery.
  • High reported purity (99.7%).
  • Stable as powder at -20°C for long-term storage.
  • Available in small pack sizes for biochemical assays.