Learn More
Medchemexpress LLC N-(6-chloro-3-pyridinyl)-3,4-difluoro-benzamide | 325457-89-4 | ≥98.0% | 10 MG
SDP
Catalog No. 5000230487
Encompass_Preferred
ICA-27243 is a selective, potent, orally active KCNQ2/KCNQ3 potassium channel opener used as a research tool to study neuronal M-currents and anticonvulsant pharmacology.
  • Selective activation of KCNQ2/KCNQ3 channels
  • Orally active with reported EC50 of 0.38 μM
  • Demonstrated antiepileptic and anticonvulsant effects in preclinical models
  • High chemical purity suitable for research use
  • Solid form, stored at -20 °C for stability
Catalog No. 50-002-30487 Supplier Medchemexpress LLC Supplier No. HY12211410MG
ICA-27243 is a selective, potent, orally active KCNQ2/KCNQ3 potassium channel opener used as a research tool to study neuronal M-currents and anticonvulsant pharmacology.
  • Selective activation of KCNQ2/KCNQ3 channels
  • Orally active with reported EC50 of 0.38 μM
  • Demonstrated antiepileptic and anticonvulsant effects in preclinical models
  • High chemical purity suitable for research use
  • Solid form, stored at -20 °C for stability