Learn More
Medchemexpress LLC N-6-Methyl-2-deoxyadenosine | 2002-35-9 | 99.8% | 265.27 | 1 ML
SDP
Catalog No. 5000452564
Encompass_Preferred
N-6-Methyl-2-deoxyadenosine is an adenine nucleoside analogue. Purine nucleoside analogs exhibit broad antitumor activity, particularly against indolent lymphoid malignancies. The anticancer mechanisms involve the inhibition of DNA synthesis and the induction of apoptosis, among other processes.
  • Adenine nucleoside analogue
  • Broad antitumor activity targeting indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis
  • For research use only
Catalog No. 50-004-52564 Supplier Medchemexpress LLC Supplier No. HYW01172510MM/1ML
N-6-Methyl-2-deoxyadenosine is an adenine nucleoside analogue. Purine nucleoside analogs exhibit broad antitumor activity, particularly against indolent lymphoid malignancies. The anticancer mechanisms involve the inhibition of DNA synthesis and the induction of apoptosis, among other processes.
  • Adenine nucleoside analogue
  • Broad antitumor activity targeting indolent lymphoid malignancies
  • Inhibits DNA synthesis
  • Induces apoptosis
  • For research use only