Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(6-Methyl-2-pyridyl)thiourea, 97%

Catalog No. AAH3388406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3388406 5 g
AAH3388403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3388406 Supplier Thermo Scientific Chemicals Supplier No. H3388406
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 49600-34-2 | C7H9N3S | 167.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49600-34-2
Molecular Formula C7H9N3S
Molecular Weight (g/mol) 167.23
MDL Number MFCD00462739
InChI Key LEWSYNYKTYXRHP-UHFFFAOYSA-N
Synonym 6-methyl-pyridin-2-yl-thiourea, 6-methylpyridin-2-yl thiourea, n-6-methylpyridin-2-yl thiourea, 1-6-methylpyridin-2-yl thiourea, thiourea, 6-methyl-2-pyridyl, n-6-methyl-2-pyridyl thiourea, n-6-methyl-2-pyridinyl thiourea #, thiourea, n-6-methyl-2-pyridinyl, thiourea,n-6-methyl-2-pyridinyl
PubChem CID 685983
IUPAC Name (6-methylpyridin-2-yl)thiourea
SMILES CC1=CC=CC(NC(N)=S)=N1

Specifications

Melting Point 193°C to 197°C
Quantity 5 g
Formula Weight 167.23
Percent Purity 97%
Chemical Name or Material N-(6-Methyl-2-pyridyl)thiourea
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.