Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-((6aS,8S)-2-[4-(methylthio)phenyl]-6,12-dioxopyrido-benzodiazepin-8-yl)-2-thiophenesulfonamide | 1193340-31-6 | 98.0% | 1 MG
SDP

Catalog No. 5000234736
Encompass_Preferred

AN-12-H5 is a research-stage antienterovirus small molecule that inhibits replication of poliovirus (PV) and enterovirus 71 (EV71). Supplied as an off-white to light yellow solid, it is accompanied by quality documentation for research use.

  • Antienterovirus activity targeting replication of PV and EV71.
  • Purity 98.0% by HPLC.
  • Molecular weight 513.65 g/mol; formula C24H23N3O4S3.
  • Available in milligram quantities with certificate of analysis and safety data sheet.
  • Off-white to light yellow solid; store refrigerated as recommended.

Catalog No. 50-002-34736 Supplier Medchemexpress LLC Supplier No. HY1209001MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AN-12-H5 is a research-stage antienterovirus small molecule that inhibits replication of poliovirus (PV) and enterovirus 71 (EV71). Supplied as an off-white to light yellow solid, it is accompanied by quality documentation for research use.

  • Antienterovirus activity targeting replication of PV and EV71.
  • Purity 98.0% by HPLC.
  • Molecular weight 513.65 g/mol; formula C24H23N3O4S3.
  • Available in milligram quantities with certificate of analysis and safety data sheet.
  • Off-white to light yellow solid; store refrigerated as recommended.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.