Learn More
Medchemexpress LLC N-(7-cyano-6-(4-fluoro-3-(2-(3-(trifluoromethyl)phenyl)acetamido)phenoxy)... | 1228591-30-7 | MFCD26960965 | >98.0% | 554.52 g/mol | C27H18F4N4O3S | 10 MG
SDP
Catalog No. 5000288002
Encompass_Preferred
TAK-632 is a potent pan-RAF kinase inhibitor supplied as a small-quantity research reagent for biochemical and cell-based studies targeting RAF signaling.
  • Potent pan-RAF inhibition: CRAF 1.4 nM; BRAF V600E 2.4 nM; BRAF WT 8.3 nM.
  • Suitable for biochemical and cell-based RAF pathway assays.
  • High purity (≥98%).
  • Provided as a small quantity solid for research use.
  • Reported molecular weight 554.52 g/mol and formula C27H18F4N4O3S.
Catalog No. 50-002-88002 Supplier Medchemexpress LLC Supplier No. HY1576710MG
TAK-632 is a potent pan-RAF kinase inhibitor supplied as a small-quantity research reagent for biochemical and cell-based studies targeting RAF signaling.
  • Potent pan-RAF inhibition: CRAF 1.4 nM; BRAF V600E 2.4 nM; BRAF WT 8.3 nM.
  • Suitable for biochemical and cell-based RAF pathway assays.
  • High purity (≥98%).
  • Provided as a small quantity solid for research use.
  • Reported molecular weight 554.52 g/mol and formula C27H18F4N4O3S.