Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(7-methoxy-4-quinolinyl)-N'-[6-(trifluoromethyl)-2-pyridinyl]-urea | 1384424-80-9 | 99.9% | 362.3 g/mol | C17H13F3N4O2 | 1 ML
SDP

Catalog No. 5000241452
Encompass_Preferred

A 1070722 is a potent and selective glycogen synthase kinase-3 (GSK-3) inhibitor (Ki = 0.6 nM) that penetrates the blood-brain barrier and has been used in PET tracer research. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for research use only.

  • Potent GSK-3 inhibition (Ki = 0.6 nM).
  • Brain-penetrant, suitable for central nervous system studies.
  • Ready-to-use solution: 10 mM in DMSO, 1 mL.
  • High purity (>99.8%).
  • Molecular weight 362.3 g/mol; formula C17H13F3N4O2.
  • Recommended storage for solution: -80°C for long-term stability.

Catalog No. 50-002-41452 Supplier Medchemexpress LLC Supplier No. HY10753110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A 1070722 is a potent and selective glycogen synthase kinase-3 (GSK-3) inhibitor (Ki = 0.6 nM) that penetrates the blood-brain barrier and has been used in PET tracer research. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for research use only.

  • Potent GSK-3 inhibition (Ki = 0.6 nM).
  • Brain-penetrant, suitable for central nervous system studies.
  • Ready-to-use solution: 10 mM in DMSO, 1 mL.
  • High purity (>99.8%).
  • Molecular weight 362.3 g/mol; formula C17H13F3N4O2.
  • Recommended storage for solution: -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.