Learn More
Medchemexpress LLC N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl)-phosphatidylethanolamine, triethylammonium salt | 178119-00-1 | MFCD00467850 | 95.0% | 956.26 g/mol | C49H90N5O11P | 10 MG
SDP
Catalog No. 5000296792
Encompass_Preferred
NBD-PE is an NBD-labeled phosphatidylethanolamine fluorescent probe used to label membranes and monitor lipid trafficking and membrane dynamics. It exhibits environment-sensitive green fluorescence (excitation ~463 nm, emission ~526 nm) and is supplied as a solid for laboratory use. CAS 178119-00-1; molecular weight ~956.26 g/mol.
  • Green fluorescent NBD label with excitation ~463 nm and emission ~526 nm.
  • Integrates into lipid bilayers for membrane labeling and tracking.
  • Environment-sensitive fluorescence suitable for monitoring membrane order.
  • Available in small laboratory quantities for experimental use.
  • Supplied as a solid with a brown to black physical appearance.
  • CAS 178119-00-1; molecular weight 956.26 g/mol.
Catalog No. 50-002-96792 Supplier Medchemexpress LLC Supplier No. HYD168310MG
NBD-PE is an NBD-labeled phosphatidylethanolamine fluorescent probe used to label membranes and monitor lipid trafficking and membrane dynamics. It exhibits environment-sensitive green fluorescence (excitation ~463 nm, emission ~526 nm) and is supplied as a solid for laboratory use. CAS 178119-00-1; molecular weight ~956.26 g/mol.
  • Green fluorescent NBD label with excitation ~463 nm and emission ~526 nm.
  • Integrates into lipid bilayers for membrane labeling and tracking.
  • Environment-sensitive fluorescence suitable for monitoring membrane order.
  • Available in small laboratory quantities for experimental use.
  • Supplied as a solid with a brown to black physical appearance.
  • CAS 178119-00-1; molecular weight 956.26 g/mol.