Learn More
Medchemexpress LLC N-(7-nitrobenzo[c][1,2,5]oxadiazol-4-yl)demethyltamoxifen | 1481401-71-1 | 99.8% | 1 ML
SDP
Catalog No. 5000234512
Encompass_Preferred
Fluorescent tamoxifen derivative used to label intracellular tamoxifen-binding sites (estrogen receptors). Supplied as a 10 mM solution in DMSO (1 mL) or as a solid; suitable for cell imaging and receptor localization studies.
  • labels intracellular tamoxifen-binding sites
  • retains antiestrogenic activity in breast cancer cells
  • devoid of estrogenic agonist effect on the uterus
  • available as a 10 mM solution in DMSO or as a solid
  • high purity suitable for biochemical and imaging assays
Catalog No. 50-002-34512 Supplier Medchemexpress LLC Supplier No. HY11943710MM 1ML
Fluorescent tamoxifen derivative used to label intracellular tamoxifen-binding sites (estrogen receptors). Supplied as a 10 mM solution in DMSO (1 mL) or as a solid; suitable for cell imaging and receptor localization studies.
  • labels intracellular tamoxifen-binding sites
  • retains antiestrogenic activity in breast cancer cells
  • devoid of estrogenic agonist effect on the uterus
  • available as a 10 mM solution in DMSO or as a solid
  • high purity suitable for biochemical and imaging assays