Learn More
Medchemexpress LLC N-(7-nitrobenzo[c][1,2,5]oxadiazol-4-yl)demethyltamoxifen | 1481401-71-1 | 99.8% | 10 MG
SDP
Catalog No. 5000234427
Encompass_Preferred
FLTX1 is a fluorescent tamoxifen derivative used to label intracellular tamoxifen-binding sites (estrogen receptors) in permeabilized and non-permeabilized cells. It retains antiestrogenic activity of tamoxifen in breast cancer cells while lacking estrogenic agonist effects on the uterus. Supplied as a high-purity research reagent for cellular imaging and receptor localization studies.
  • Fluorescent probe for estrogen receptor localization.
  • Retains antiestrogenic activity without uterine agonism.
  • High purity suitable for imaging and biochemical assays.
  • Soluble in DMSO; available as solid or solution formats.
  • Available in multiple packaging sizes, including 10 MG for small-scale studies.
Catalog No. 50-002-34427 Supplier Medchemexpress LLC Supplier No. HY11943710MG
FLTX1 is a fluorescent tamoxifen derivative used to label intracellular tamoxifen-binding sites (estrogen receptors) in permeabilized and non-permeabilized cells. It retains antiestrogenic activity of tamoxifen in breast cancer cells while lacking estrogenic agonist effects on the uterus. Supplied as a high-purity research reagent for cellular imaging and receptor localization studies.
  • Fluorescent probe for estrogen receptor localization.
  • Retains antiestrogenic activity without uterine agonism.
  • High purity suitable for imaging and biochemical assays.
  • Soluble in DMSO; available as solid or solution formats.
  • Available in multiple packaging sizes, including 10 MG for small-scale studies.