Learn More
Cayman Chemical N-(8Z-Heptadecen-1-yl)-O-(3-pyridylmethyl)carbamate, Purity: ≥98%, CAS: 1450603-63-0, 100 mg
SDP
Catalog No. 502892790
Encompass_Preferred
A synthetic macamide that irreversibly inhibits FAAH with an IC50 value of 0.153 µM
Catalog No. 50-289-2790 Supplier Cayman Chemical Supplier No. 9002365100 MG
A synthetic macamide that irreversibly inhibits FAAH with an IC50 value of 0.153 µM