Learn More
Cayman Chemical N 8ZHeptadecen1ylO 3p 10mg
SDP
Catalog No. 502893048
Encompass_Preferred
A synthetic macamide that irreversibly inhibits FAAH with an IC50 value of 0.153 µM
Catalog No. 50-289-3048 Supplier Cayman Chemical Supplier No. 900236510 MG
A synthetic macamide that irreversibly inhibits FAAH with an IC50 value of 0.153 µM