Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(9-fluorenylmethoxycarbonyl)-L-leucine (13C-labeled) | 202114-53-2 | MFCD00190328 | 98.0% | 354.4 g/mol | C20(13C)H23NO4 | 10 MG
SDP

Catalog No. 5000212149
Encompass_Preferred

Fmoc-leucine-13C is an isotopically labeled N-(9-fluorenylmethoxycarbonyl)-L-leucine used as a protected, 13C-labeled amino acid building block for peptide synthesis and analytical applications. It provides a stable isotope tracer for mass spectrometry and structural studies, with high chemical purity suitable for research use.

  • Isotopically labeled with 13C for tracing and mass spectrometry.
  • Fmoc-protected L-leucine suitable for solid-phase peptide synthesis.
  • High chemical purity (≈98% by HPLC).
  • Molecular weight 354.4 g/mol and formula C20(13C)H23NO4.
  • Available in small research quantities for analytical standards and synthesis.

Catalog No. 50-002-12149 Supplier Medchemexpress LLC Supplier No. HY101064S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-leucine-13C is an isotopically labeled N-(9-fluorenylmethoxycarbonyl)-L-leucine used as a protected, 13C-labeled amino acid building block for peptide synthesis and analytical applications. It provides a stable isotope tracer for mass spectrometry and structural studies, with high chemical purity suitable for research use.

  • Isotopically labeled with 13C for tracing and mass spectrometry.
  • Fmoc-protected L-leucine suitable for solid-phase peptide synthesis.
  • High chemical purity (≈98% by HPLC).
  • Molecular weight 354.4 g/mol and formula C20(13C)H23NO4.
  • Available in small research quantities for analytical standards and synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.