Learn More
Medchemexpress LLC N-(((9H-fluoren-9-yl)methoxy)carbonyl)-N-(2,4-dimethoxybenzyl)glycine | 166881-42-1 | MFCD08064316 | 99.7% | 447.49 g/mol | C26H25NO6 | 5 G
SDP
Catalog No. 5000210883
Encompass_Preferred
N-(((9H-fluoren-9-yl)methoxy)carbonyl)-N-(2,4-dimethoxybenzyl)glycine is an Fmoc-protected glycine derivative used as a protected amino-acid building block in peptide synthesis and related organic synthesis. It has molecular formula C26H25NO6 and molecular weight 447.49 g/mol.
  • Fmoc-protected glycine derivative suitable for solid-phase peptide synthesis.
  • High purity appropriate for research use.
  • Available in small laboratory pack sizes for convenience.
  • Molecular formula C26H25NO6 and molecular weight 447.49 g/mol aid in formulation and calculation.
Catalog No. 50-002-10883 Supplier Medchemexpress LLC Supplier No. HYW0477945G
N-(((9H-fluoren-9-yl)methoxy)carbonyl)-N-(2,4-dimethoxybenzyl)glycine is an Fmoc-protected glycine derivative used as a protected amino-acid building block in peptide synthesis and related organic synthesis. It has molecular formula C26H25NO6 and molecular weight 447.49 g/mol.
  • Fmoc-protected glycine derivative suitable for solid-phase peptide synthesis.
  • High purity appropriate for research use.
  • Available in small laboratory pack sizes for convenience.
  • Molecular formula C26H25NO6 and molecular weight 447.49 g/mol aid in formulation and calculation.