Learn More
Medchemexpress LLC N-(((9H-fluoren-9-yl)methoxy)carbonyl)-N-allylglycine | 222725-35-1 | 97.0% | 337.38 g·mol⁻¹ | C20H19NO4 | 100 MG
SDP
Catalog No. 5000301190
Encompass_Preferred
N-(((9H-fluoren-9-yl)methoxy)carbonyl)-N-allylglycine is an Fmoc-protected N-allylglycine derivative used as a protected amino acid building block in solid-phase peptide synthesis and related organic synthesis. The Fmoc group protects the amino function while the N-allyl substituent enables orthogonal protection strategies and selective transformations.
  • Fmoc-protected amino acid for peptide synthesis.
  • N-allyl substituent allows orthogonal deprotection and selective modification.
  • Suitable for standard peptide coupling reagents and conditions.
  • Supports orthogonal synthetic strategies in complex sequences.
  • Supplied in research-scale quantities for laboratory use.
Catalog No. 50-003-01190 Supplier Medchemexpress LLC Supplier No. HYW036352100MG
N-(((9H-fluoren-9-yl)methoxy)carbonyl)-N-allylglycine is an Fmoc-protected N-allylglycine derivative used as a protected amino acid building block in solid-phase peptide synthesis and related organic synthesis. The Fmoc group protects the amino function while the N-allyl substituent enables orthogonal protection strategies and selective transformations.
  • Fmoc-protected amino acid for peptide synthesis.
  • N-allyl substituent allows orthogonal deprotection and selective modification.
  • Suitable for standard peptide coupling reagents and conditions.
  • Supports orthogonal synthetic strategies in complex sequences.
  • Supplied in research-scale quantities for laboratory use.