Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(((9H-Fluoren-9-yl)methoxy)carbonyl)-N-allylglycine | 222725-35-1 | MFCD17976766 | 337.38 g·mol⁻¹ | C20H19NO4 | 1 G
SDP

Catalog No. 5000299722
Encompass_Preferred

N-(((9H-Fluoren-9-yl)methoxy)carbonyl)-N-allylglycine is an Fmoc-protected amino acid derivative provided as a powder for laboratory synthesis. It is used as a protected glycine building block in peptide synthesis, method development, and other organic chemistry applications where temporary N-protection and allyl functionality are required.

  • Powder physical form for convenient storage and handling.
  • Molecular formula C20H19NO4 and molecular weight 337.38 g·mol⁻¹.
  • Suitable as an Fmoc-protected glycine derivative for peptide coupling.
  • Allyl functionality allows for selective deprotection or modification.
  • Storage recommendations include -20°C for long-term stability.
  • Offered in small research quantities for bench-scale synthesis.

Catalog No. 50-002-99722 Supplier Medchemexpress LLC Supplier No. HYW0363521G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-(((9H-Fluoren-9-yl)methoxy)carbonyl)-N-allylglycine is an Fmoc-protected amino acid derivative provided as a powder for laboratory synthesis. It is used as a protected glycine building block in peptide synthesis, method development, and other organic chemistry applications where temporary N-protection and allyl functionality are required.

  • Powder physical form for convenient storage and handling.
  • Molecular formula C20H19NO4 and molecular weight 337.38 g·mol⁻¹.
  • Suitable as an Fmoc-protected glycine derivative for peptide coupling.
  • Allyl functionality allows for selective deprotection or modification.
  • Storage recommendations include -20°C for long-term stability.
  • Offered in small research quantities for bench-scale synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.