Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(((9H-fluoren-9-yl)methoxy)carbonyl)-O-trityl-L-homoserine | 111061-55-3 | MFCD00270544 | 97.0% | 583.67 g/mol | C38H33NO5 | 10 G
SDP

Catalog No. 5000210888
Encompass_Preferred

N-(((9H-fluoren-9-yl)methoxy)carbonyl)-O-trityl-L-homoserine is an Fmoc- and O-trityl-protected L-homoserine derivative used as a building block in peptide synthesis and amino-acid chemistry. It is supplied with high purity for research applications.

  • Fmoc- and O-trityl-protected L-homoserine used for peptide synthesis.
  • CAS number 111061-55-3 for unambiguous identification.
  • Molecular formula C38H33NO5 and molecular weight 583.67 g/mol.
  • Typical purity about 97.0% as supplied.
  • Soluble at 10 mM in DMSO for stock solutions.
  • Packaged as a 10 g quantity suitable for synthetic workflows.

Catalog No. 50-002-10888 Supplier Medchemexpress LLC Supplier No. HYW04866810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-(((9H-fluoren-9-yl)methoxy)carbonyl)-O-trityl-L-homoserine is an Fmoc- and O-trityl-protected L-homoserine derivative used as a building block in peptide synthesis and amino-acid chemistry. It is supplied with high purity for research applications.

  • Fmoc- and O-trityl-protected L-homoserine used for peptide synthesis.
  • CAS number 111061-55-3 for unambiguous identification.
  • Molecular formula C38H33NO5 and molecular weight 583.67 g/mol.
  • Typical purity about 97.0% as supplied.
  • Soluble at 10 mM in DMSO for stock solutions.
  • Packaged as a 10 g quantity suitable for synthetic workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.